Please login to your online account to display your discounted pricing

Alfa Aesar™ Thiotriazinone, 98+%

$74.51 - $972.82

Chemical Identifiers

Chemical Name or Material Thiotriazinone
CAS 58909-39-0
Molecular Formula C4H5N3O2S
Formula Weight 159.16
MDL Number MFCD00185756
Synonym Tetrahydro-2-methyl-3-thioxo-1,2,4-triazine-5,6-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2266309

 
alfa aesar™
B2266309
10g Each for $74.51

AAB2266318

 
alfa aesar™
B2266318
50g Each for $267.84

AAB2266330

 
alfa aesar™
B2266330
250g Each for $972.82
Description & Specifications

Specifications

Formula Weight 159.16
Chemical Name or Material Thiotriazinone
Assay ≥98%
Molecular Formula C4H5N3O2S
MDL Number MFCD00185756
CAS 58909-39-0
Synonym Tetrahydro-2-methyl-3-thioxo-1,2,4-triazine-5,6-dione
Melting Point 168° to 171°C
Quantity 10g