Learn More
Thiosemicarbazide, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 138905000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Thiosemicarbazide | |
79-19-6 | |
White to Yellow | |
98+% | |
CH5N3S | |
MFCD00007620 | |
03,195 | |
15,9514 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol | |
C(=S)(N)NN | |
91.13 | |
91.13 | |
Powder |
99% | |
175.0°C to 180.0°C | |
Authentic | |
Plastic bottle | |
H2NCSNHNH2 | |
500 g | |
01,1164 | |
thiosemicarbazide, hydrazinecarbothioamide, n-aminothiourea, 1-aminothiourea, semicarbazide, thio, isothiosemicarbazide, 2-thiosemicarbazide, 3-thiosemicarbazide, thiocarbamylhydrazine, thiocarbamoylhydrazine | |
BRWIZMBXBAOCCF-UHFFFAOYSA-N | |
aminothiourea | |
2723789 | |
98+% |
Chemical Identifiers
79-19-6 | |
91.13 | |
BRWIZMBXBAOCCF-UHFFFAOYSA-N | |
2723789 | |
C(=S)(N)NN |
CH5N3S | |
MFCD00007620 | |
thiosemicarbazide, hydrazinecarbothioamide, n-aminothiourea, 1-aminothiourea, semicarbazide, thio, isothiosemicarbazide, 2-thiosemicarbazide, 3-thiosemicarbazide, thiocarbamylhydrazine, thiocarbamoylhydrazine | |
aminothiourea |
Safety and Handling
GHS H Statement
Fatal if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not eat,drink or smoke when using this product.
GHS Signal Word: Danger
EINECSNumber : 201-184-7
RTECSNumber : VT4200000
TSCA : TSCA
RUO – Research Use Only