Learn More
Thiosalicylic acid, 98%, Thermo Scientific Chemicals
$147.64 - $1351.58
Chemical Identifiers
CAS | 147-93-3 |
---|---|
Molecular Formula | C7H6O2S |
Molecular Weight (g/mol) | 154.183 |
MDL Number | MFCD00004836 |
InChI Key | NBOMNTLFRHMDEZ-UHFFFAOYSA-N |
Synonym | thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto |
PubChem CID | 5443 |
ChEBI | CHEBI:59124 |
IUPAC Name | 2-sulfanylbenzoic acid |
SMILES | C1=CC=C(C(=C1)C(=O)O)S |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1340118
|
Thermo Scientific Chemicals
A1340118 |
50 g |
Each for $147.64
|
|
|||||
AAA1340130
|
Thermo Scientific Chemicals
A1340130 |
250 g |
Each for $478.85
|
|
|||||
AAA134010B
|
Thermo Scientific Chemicals
A134010B |
1000 g |
Each for $1,351.58
|
|
|||||
Description
Thiosalicylic acid acts as a trapping agent, which is useful in the desulfenylation of 3-indolyl sulfides. It is used to prepare macrocyclic diamides by condensation with diamines. Further, it is used as a precursor to drug, which finds application for the treatment of atherosclerosis and melanoma. In addition to this, it is involved in the preparation of vaccine preservative thiomersal and thioindigo dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThiosalicylic acid acts as a trapping agent, which is useful in the desulfenylation of 3-indolyl sulfides. It is used to prepare macrocyclic diamides by condensation with diamines. Further, it is used as a precursor to drug, which finds application for the treatment of atherosclerosis and melanoma. In addition to this, it is involved in the preparation of vaccine preservative thiomersal and thioindigo dyestuff.
Solubility
Soluble in dimethyl sulfoxide, alcohol and ether. Slightly soluble in water, ethanol and alkanes.
Notes
Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
147-93-3 | |
154.183 | |
NBOMNTLFRHMDEZ-UHFFFAOYSA-N | |
5443 | |
2-sulfanylbenzoic acid |
C7H6O2S | |
MFCD00004836 | |
thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto | |
CHEBI:59124 | |
C1=CC=C(C(=C1)C(=O)O)S |
Specifications
165°C to 168°C | |
C7H6O2S | |
149360 | |
Soluble in dimethyl sulfoxide,alcohol and ether. Slightly soluble in water,ethanol and alkanes. | |
C1=CC=C(C(=C1)C(=O)O)S | |
154.183 | |
CHEBI:59124 | |
98% | |
50 g | |
Air and light sensitive |
147-93-3 | |
MFCD00004836 | |
thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto | |
NBOMNTLFRHMDEZ-UHFFFAOYSA-N | |
2-sulfanylbenzoic acid | |
5443 | |
154.19 | |
1.49 | |
Stench | |
Thiosalicylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-704-3
RTECSNumber : DH3325000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only