Learn More
Thiophosgene, 85%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 151640250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
73.0°C | |
463-71-8 | |
Liquid | |
80% min. (GC) | |
ClCSCl | |
03,134 | |
62°C | |
Solubility in water: slow decomposition in cold, fast in hot water. Other solubilities: soluble in ether, ccl4, decomposes in alcohol | |
C(=S)(Cl)Cl | |
114.98 | |
CHEBI:29366 | |
85% | |
1.5442 | |
1.5080g/mL |
Thiophosgene | |
Orange-Red to Red | |
25 g | |
CCl2S | |
MFCD00004918 | |
05,667; 09,466 | |
thiophosgene, carbonothioic dichloride, carbon chlorosulfide, thiocarbonyl chloride, thiocarbonic dichloride, dichlorothiocarbonyl, thiofosgen, phosgene, thio, thiokarbonylchlorid, carbonyl chloride, thio | |
ZWZVWGITAAIFPS-UHFFFAOYSA-N | |
thiocarbonyl dichloride | |
10040 | |
114.98 | |
Glass bottle | |
1.508 |
Chemical Identifiers
463-71-8 | |
ZWZVWGITAAIFPS-UHFFFAOYSA-N | |
10040 | |
thiocarbonyl dichloride |
MFCD00004918 | |
thiophosgene, carbonothioic dichloride, carbon chlorosulfide, thiocarbonyl chloride, thiocarbonic dichloride, dichlorothiocarbonyl, thiofosgen, phosgene, thio, thiokarbonylchlorid, carbonyl chloride, thio | |
CHEBI:29366 | |
C(=S)(Cl)Cl |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
If skin i
GHS Signal Word: Danger
EINECSNumber : 207-341-6
RUO – Research Use Only