Learn More
Thiophenol, 99%, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 430721000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Thiophenol | |
-15.0°C | |
1.0780g/mL | |
50°C | |
98.5% min. (GC) | |
C6H6S | |
C6H5SH | |
100 mL | |
15,9508 | |
Solubility in water: insoluble. Other solubilities: very soluble in alcohol, miscible with ether, benzene and cs2 | |
C1=CC=C(C=C1)S | |
110.17 | |
0.981 mm2/s (30°C) | |
110.17 | |
Liquid |
108-98-5 | |
Colorless to Yellow | |
167.0°C to 169.0°C | |
Authentic | |
AcroSeal™ Glass bottle | |
1.5880 to 1.5900 | |
MFCD00004826 | |
1.078 | |
thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto | |
RMVRSNDYEFQCLF-UHFFFAOYSA-N | |
benzenethiol | |
7969 | |
CHEBI:48498 | |
99% |
Chemical Identifiers
108-98-5 | |
110.17 | |
RMVRSNDYEFQCLF-UHFFFAOYSA-N | |
7969 | |
benzenethiol |
C6H6S | |
MFCD00004826 | |
thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto | |
CHEBI:48498 | |
C1=CC=C(C=C1)S |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May cause respiratory irritation.
Causes skin irritation.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Suspected of damaging fertility or the unbo
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thorou
GHS Signal Word: Danger
EINECSNumber : 203-635-3
RUO – Research Use Only