Learn More
Thiophenol, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1591618
Description
Thiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.
Solubility
Miscible with most organic solvents and aqueous base. Immiscible with water.
Notes
Incompatible with strong acids, bases and oxidizing agents.
Specifications
Thiophenol | |
-15°C | |
5 | |
55°C (131°F) | |
Stench | |
1.59 | |
50 g | |
506523 | |
14,9355 | |
Miscible with most organic solvents and aqueous base. Immiscible with water. | |
C1=CC=C(C=C1)S | |
110.174 | |
CHEBI:48498 | |
≥99% |
108-98-5 | |
1.075 | |
168°C to 169°C | |
≥99% | |
C6H6S | |
MFCD00004826 | |
UN2337 | |
Air Sensitive | |
thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto | |
RMVRSNDYEFQCLF-UHFFFAOYSA-N | |
benzenethiol | |
7969 | |
110.18 | |
Liquid |
Chemical Identifiers
108-98-5 | |
110.174 | |
RMVRSNDYEFQCLF-UHFFFAOYSA-N | |
7969 | |
benzenethiol |
C6H6S | |
MFCD00004826 | |
thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercapto | |
CHEBI:48498 | |
C1=CC=C(C=C1)S |
Safety and Handling
GHS H Statement
H300-H310-H330-H318-H226-H361-H315-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
May cause respiratory irritation.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280i-P281-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P330-P332+P313-P363-P37
H226-H300+H310+H330-H315-H319-H335-H361-H371-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: PHENYL MERCAPTAN
EINECSNumber : 203-635-3
RTECSNumber : DC0525000
TSCA : Yes
storageNote1 : Air sensitive
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only