Learn More
Thiophene, 99+%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 138821000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Thiophene | |
110-02-1 | |
1.0650g/mL | |
−9°C | |
99% min. (GC) | |
C4H4S | |
MFCD00005413 | |
17, 29 | |
15, 9506 | |
Solubility in water: insoluble. Other solubilities: miscible with most organic solvents | |
S1C=CC=C1 | |
84.14 | |
CHEBI:30856 | |
99+% | |
Liquid |
99+% | |
-38.0°C | |
84.0°C | |
Authentic | |
Glass bottle | |
1.5270 to 1.5290 | |
100 g | |
1.065 | |
thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio | |
YTPLMLYBLZKORZ-UHFFFAOYSA-N | |
thiophene | |
8030 | |
84.14 | |
Extra Pure |
Chemical Identifiers
110-02-1 | |
84.14 | |
YTPLMLYBLZKORZ-UHFFFAOYSA-N | |
8030 | |
thiophene |
C4H4S | |
MFCD00005413 | |
thiofuran, thiole, thiophen, thiotetrole, thiacyclopentadiene, divinylene sulfide, thiaphene, thiofuram, thiofurfuran, furan, thio | |
CHEBI:30856 | |
S1C=CC=C1 |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Harmful if swallowed.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
EINECSNumber : 203-729-4
RTECSNumber : XM7350000
TSCA : TSCA
RUO – Research Use Only