Please login to your online account to display your discounted pricing

Alfa Aesar™ Thiophene-3-carboxaldoxime, 98%

$162.73 - $162.73

Chemical Identifiers

Chemical Name or Material Thiophene-3-carboxaldoxime
CAS 42466-50-2
Molecular Formula C5H5NOS
Formula Weight 127.17
MDL Number MFCD00151853
Beilstein 108818
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1311406

  SDS
alfa aesar™
L1311406
5g Each for $162.73
Description & Specifications

Specifications

Formula Weight 127.17
Chemical Name or Material Thiophene-3-carboxaldoxime
Assay 98%
MDL Number MFCD00151853
CAS 42466-50-2
Molecular Formula C5H5NOS
Quantity 5g
Melting Point 121° to 122°C
Beilstein 108818