Please login to your online account to display your discounted pricing

Alfa Aesar™ Thiophene-3-carboxaldehyde diethyl acetal, 98%

$57.09 - $183.18

Chemical Identifiers

Chemical Name or Material Thiophene-3-carboxaldehyde diethyl acetal
CAS 3199-44-8
Molecular Formula C9H14O2S
Formula Weight 186.27
MDL Number MFCD01318173
Beilstein 125429
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1552006

 
alfa aesar™
L1552006
5g Each for $183.18

AAL1552003

 
alfa aesar™
L1552003
1g Each for $57.09
Description & Specifications

Specifications

Formula Weight 186.27
Chemical Name or Material Thiophene-3-carboxaldehyde diethyl acetal
Assay 98%
Molecular Formula C9H14O2S
MDL Number MFCD01318173
CAS 3199-44-8
Beilstein 125429
Boiling Point 130°C (30mmHg)
Quantity 5g

Applications
Thiophene-3-carboxaldehyde diethyl aceta is used in OLED materials. Also used as electronic intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.