missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Thiophene-3-carbonitrile, 98%

Manufacturer:  Alfa Aesar™ L1311514

 View more versions of this product

Catalog No. AAL1311514



Specifications

Specifications

Thiophene-3-carbonitrile
Recognizable
25g
C5H3NS
UN3276
3-cyanothiophene, 3-thiophenecarbonitrile, thiophene-3-nitrile, pubchem5508, pubchem12351, acmc-209dqk, ksc176g4h, 3-thiophenecarbonitrile
C1=CSC=C1C#N
109.146
109.15
86°C (186°F)
205°C
1641-09-4
MFCD00151852
107104
GSXCEVHRIVLFJV-UHFFFAOYSA-N
thiophene-3-carbonitrile
74231
98%

Chemical Identifiers

1641-09-4
109.146
GSXCEVHRIVLFJV-UHFFFAOYSA-N
74231
C1=CSC=C1C#N
C5H3NS
MFCD00151852
3-cyanothiophene, 3-thiophenecarbonitrile, thiophene-3-nitrile, pubchem5508, pubchem12351, acmc-209dqk, ksc176g4h, 3-thiophenecarbonitrile
thiophene-3-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H312-H332-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

Documents