Please login to your online account to display your discounted pricing

Alfa Aesar™ Thiophene-2-thiocarboxamide, 97%

Chemical Identifiers

Chemical Name or Material Thiophene-2-thiocarboxamide
CAS 20300-02-1
Molecular Formula C5H5NS2
Formula Weight 143.23
MDL Number MFCD00052890
UN Number UN2811
Beilstein 110159
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2522506

 
alfa aesar™
B2522506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2522514

 
alfa aesar™
B2522514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 143.23
Chemical Name or Material Thiophene-2-thiocarboxamide
Assay 97%
Molecular Formula C5H5NS2
MDL Number MFCD00052890
CAS 20300-02-1
UN Number UN2811
Beilstein 110159
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Melting Point 106° to 109°C
Quantity 5g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Applications
Thiophene-2-thiocarboxamide is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.