missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiophene-2-ethylamine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC354270050

 View more versions of this product

Catalog No. AC354270050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

127.21
≥97.5%
Colorless to yellow
Authentic
C6H9NS
98%
30433-91-1
Liquid
5g
1.5500 to 1.5530 (20°C, 589nm)
Glass bottle
Thiophene-2-ethylamine
MFCD00051495

Chemical Identifiers

30433-91-1
C6H9NS
MFCD00051495
≥97.5%
98%
127.21
Thiophene-2-ethylamine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 250-196-9