missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Thiophene-2-carboxylic hydrazide, 98+%

$64.62 - $644.74

Chemical Identifiers

CAS 2361-27-5
Molecular Formula C5H6N2OS
Formula Weight 142.18
MDL Number MFCD00005435
Beilstein 114437
Chemical Name or Material Thiophene-2-carboxylic hydrazide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1127006 SDS Alfa Aesar™
A1127006
5g Each for $64.62
AAA1127014 SDS Alfa Aesar™
A1127014
25g Each for $194.47
AAA1127022 SDS Alfa Aesar™
A1127022
100g Each for $644.74
Specifications Description & Specifications

Chemical Identifiers

2361-27-5
142.18
114437
C5H6N2OS
MFCD00005435
Thiophene-2-carboxylic hydrazide

Specifications

2361-27-5
142.18
114437
5g
≥98%
C5H6N2OS
MFCD00005435
137° to 140°C
Thiophene-2-carboxylic hydrazide

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning