missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Thiophene-2-carboxylic anhydride, 95%

Manufacturer:  Alfa Aesar™ L1285103

 View more versions of this product

Catalog No. AAL1285103



Description

Description

Applications
Thiophene-2-carboxylic anhydride is used as an organic chemical synthesis intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

Thiophene-2-carboxylic anhydride
1g
C10H6O3S2
177049
BKBCDDYQJQGMNX-UHFFFAOYSA-N
thiophene-2-carbonyl thiophene-2-carboxylate
3680768
95%
58°C to 61°C
25569-97-5
MFCD00068089
thiophene-2-carboxylic anhydride, thiophene-2-carboxylic acid anhydride, 2-thenoyl ether, 2-thenoic anhydride, acmc-20ajil, bis 2-thiophenecarboxylic acid anhydride, 2-thiophenecarboxylicacid,1,1'-anhydride, 2-thiophenecarboxylicacid, 1,1'-anhydride, thiophen-2-ylcarbonyl thiophene-2-carboxylate, 2-thiophenecarboxylic acid oxo thiophen-2-yl methyl ester
C1=CSC(=C1)C(=O)OC(=O)C2=CC=CS2
238.275
238.29

Chemical Identifiers

25569-97-5
238.275
BKBCDDYQJQGMNX-UHFFFAOYSA-N
3680768
C1=CSC(=C1)C(=O)OC(=O)C2=CC=CS2
C10H6O3S2
MFCD00068089
thiophene-2-carboxylic anhydride, thiophene-2-carboxylic acid anhydride, 2-thenoyl ether, 2-thenoic anhydride, acmc-20ajil, bis 2-thiophenecarboxylic acid anhydride, 2-thiophenecarboxylicacid,1,1'-anhydride, 2-thiophenecarboxylicacid, 1,1'-anhydride, thiophen-2-ylcarbonyl thiophene-2-carboxylate, 2-thiophenecarboxylic acid oxo thiophen-2-yl methyl ester
thiophene-2-carbonyl thiophene-2-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents