Please login to your online account to display your discounted pricing

Thiophene-2-carboximidamide hydrochloride, ≥95%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RF01040DA

 View more versions of this product

Catalog No. RF01040DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 162.64
Quantity 1g
Assay 95%
CAS Min % 95.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
Molecular Formula C5H6N2S·ClH
Chemical Name or Material 2-Thiophenecarboximidamide hydrochloride
Packaging Amber Glass Bottle
MDL Number MFCD00173786
CAS 54610-70-7

Specifications

Chemical Name or Material 2-Thiophenecarboximidamide hydrochloride
CAS 54610-70-7
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C5H6N2S·ClH
Formula Weight 162.64
MDL Number MFCD00173786
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents