missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiophene-2-carboximidamide hydrochloride, ≥95%, Maybridge

$125.97 - $125.97

Chemical Identifiers

CAS 54610-70-7
CAS Min % 95.0
Assay Percent Range 95%
Molecular Formula C5H6N2S·ClH
Formula Weight 162.64
MDL Number MFCD00173786
Chemical Name or Material 2-Thiophenecarboximidamide hydrochloride
Assay 95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
RF01040DA SDS Fisher Scientific Company
RF01040DA
Amber Glass Bottle 1g Each for $125.97
Specifications Description & Specifications

Chemical Identifiers

54610-70-7
95%
162.64
2-Thiophenecarboximidamide hydrochloride
95.0
C5H6N2S·ClH
MFCD00173786
95%

Specifications

162.64
95.0
C5H6N2S·ClH
54610-70-7
2-Thiophenecarboximidamide hydrochloride
1g
95%
MFCD00173786
Amber Glass Bottle
95%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.