missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thiophene-2-carboxamide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1521914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Thiophene-2-carboxamide | |
178°C to 183°C | |
MFCD00010425 | |
110153 | |
DENPQNAWGQXKCU-UHFFFAOYSA-N | |
thiophene-2-carboxamide | |
22063 | |
99% |
5813-89-8 | |
C5H5NOS | |
25 g | |
2-thiophenecarboxamide, 2-thienylamide, 2-thenamide, thiophen-2-carboxamide, 2-aminocarbonyl thiophene, 2-thienylamide french, thiophenecarboxamide, thiolamide, acmc-1ao1m, 5-18-06-00162 beilstein handbook reference | |
C1=CSC(=C1)C(=O)N | |
127.161 | |
127.17 |
Chemical Identifiers
5813-89-8 | |
127.161 | |
DENPQNAWGQXKCU-UHFFFAOYSA-N | |
22063 | |
C1=CSC(=C1)C(=O)N |
C5H5NOS | |
MFCD00010425 | |
2-thiophenecarboxamide, 2-thienylamide, 2-thenamide, thiophen-2-carboxamide, 2-aminocarbonyl thiophene, 2-thienylamide french, thiophenecarboxamide, thiolamide, acmc-1ao1m, 5-18-06-00162 beilstein handbook reference | |
thiophene-2-carboxamide |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : XM8290000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only