Please login to your online account to display your discounted pricing

Thioperamide, Tocris


H3 antagonist and H4 inverse agonist

Quantity 10mg

Manufacturer: r & d systems  RDS064410

 View more versions of this product

Catalog No. 064410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 292.44
Molecular Formula C15H24N4S
CAS 106243-16-7
Synonym N-Cyclohexyl-4-(imidazol-4-yl)-1-piperidinecarbothioamide
Chemical Name or Material Thioperamide
Assay Percent Range >99%
Recommended Storage Store at -20°C

Specifications

Chemical Name or Material Thioperamide
Assay Percent Range >99%
Molecular Formula C15H24N4S
Formula Weight 292.44
Synonym N-Cyclohexyl-4-(imidazol-4-yl)-1-piperidinecarbothioamide
Accessories

Store at -20°C