Please login to your online account to display your discounted pricing

Thioperamide, Tocris

H3 antagonist and H4 inverse agonist

$163.07 - $678.85

Chemical Identifiers

Chemical Name or Material Thioperamide
Assay Percent Range >99%
Molecular Formula C15H24N4S
Formula Weight 292.44
Synonym N-Cyclohexyl-4-(imidazol-4-yl)-1-piperidinecarbothioamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

064450

 
r & d systems
RDS064450
50mg Each for $678.85

064410

 
r & d systems
RDS064410
10mg Each for $163.07
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 292.44
Chemical Name or Material Thioperamide
Assay Percent Range >99%
Molecular Formula C15H24N4S
CAS 106243-16-7
Synonym N-Cyclohexyl-4-(imidazol-4-yl)-1-piperidinecarbothioamide
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C