missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thionin acetate, Thermo Scientific Chemicals

Used as a biogical stain

Supplier:  Thermo Scientific Chemicals A1891206

 View more versions of this product

Catalog No. AAA1891206



Description

Description

Thionin acetate is a strongly staining metachromatic dye that is widely used for biological staining. Thionine can also be used in place of Schiff reagent in quantitative Feulgen staining of DNA. It can also be used to mediate electron transfer in microbial fuel cells. It is also employed as a general nuclear stain for chromatin and mucin; for staining Nissl granules by end-point buffering and for showing malignant cells in frozen biopsy sections; and as a stain for semi-thin sections of plant material for light microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thionin acetate is a strongly staining metachromatic dye that is widely used for biological staining. Thionine can also be used in place of Schiff reagent in quantitative Feulgen staining of DNA. It can also be used to mediate electron transfer in microbial fuel cells. It is also employed as a general nuclear stain for chromatin and mucin; for staining Nissl granules by end-point buffering and for showing malignant cells in frozen biopsy sections; and as a stain for semi-thin sections of plant material for light microscopy.

Solubility
Soluble in water (∼2.5 mg/ml at 25°C), and HOAc: H2O (1:1) (1 mg/ml). Insoluble in ethanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

Thionin
C14H13N3O2S
Soluble in water (∽2.5mg/ml at 25°C),and HOAc: H2O (1:1) (1mg/ml). Insoluble in ethanol.
CC(=O)[O-].C1=CC2=C(C=C1N)SC3=CC(=[NH2+])C=CC3=N2
287.337
287.34
78338-22-4
MFCD00081194
JOIRQYHDJINFGA-UHFFFAOYSA-N
(7-aminophenothiazin-3-ylidene)azanium;acetate
2724414
5 g

Chemical Identifiers

78338-22-4
287.337
JOIRQYHDJINFGA-UHFFFAOYSA-N
(7-aminophenothiazin-3-ylidene)azanium;acetate
C14H13N3O2S
MFCD00081194
2724414
CC(=O)[O-].C1=CC2=C(C=C1N)SC3=CC(=[NH2+])C=CC3=N2
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only