Please login to your online account to display your discounted pricing

Thionicotinamide 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  138800250

 View more versions of this product

Catalog No. AC138800250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Powder
Formula Weight 138.19
Melting Point 183° to 190°C (dec.)
Quantity 25g
Color Yellow to yellow-green
EINECS Number 225-036-6
TSCA TSCA
RTECS Number QS4488000
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Chemical Name or Material Thionicotinamide
Assay Percent Range 99%
Molecular Formula C6H6N2S
MDL Number MFCD00006399
CAS 4621-66-3

Specifications

Chemical Name or Material Thionicotinamide
Name Note 99%
CAS 4621-66-3
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H6N2S
Formula Weight 138.19
MDL Number MFCD00006399