Please login to your online account to display your discounted pricing

Alfa Aesar™ Thiomorpholine, polymer-supported, 0.8-1.1mmol/g on Merrifield resin

Chemical Identifiers

Chemical Name or Material Thiomorpholine
Name Note polymer-supported, 0.8 to 1.1mmol/g on Merrifield resin
MDL Number MFCD03458715
Synonym Methylthiomorpholine on polystyrene; 4-(Thiomorpholine)methyl, polymer-supported
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1947006

 
alfa aesar™
L1947006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1947014

 
alfa aesar™
L1947014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Thiomorpholine
MDL Number MFCD03458715
Name Note polymer-supported, 0.8 to 1.1mmol/g on Merrifield resin
Synonym Methylthiomorpholine on polystyrene; 4-(Thiomorpholine)methyl, polymer-supported
Quantity 5g