missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiomalic Acid 98.0+%, TCI America™
Supplier: TCI America M0064100G
Specifications
| Thiomalic Acid | |
| 152°C | |
| C4H6O4S | |
| 100 g | |
| NJRXVEJTAYWCQJ-UHFFFAOYSA-N | |
| 2-sulfanylbutanedioic acid | |
| 6268 | |
| 150.15 | |
| Crystalline Powder |
| 70-49-5 | |
| White-Yellow | |
| MFCD00004860 | |
| mercaptosuccinic acid, thiomalic acid, 2-thiomalic acid, 2-mercaptosuccinic acid, monomercaptosuccinic acid, butanedioic acid, mercapto, malic acid, 2-thio, mercaptosuccinate, 2-sulfanylsuccinic acid, dl-mercaptosuccinic acid | |
| C(C(C(=O)O)S)C(=O)O | |
| 150.148 | |
| CHEBI:38705 | |
| ≥98.0% (T) |
Chemical Identifiers
| 70-49-5 | |
| 150.148 | |
| NJRXVEJTAYWCQJ-UHFFFAOYSA-N | |
| 6268 | |
| 2-sulfanylbutanedioic acid |
| C4H6O4S | |
| MFCD00004860 | |
| mercaptosuccinic acid, thiomalic acid, 2-thiomalic acid, 2-mercaptosuccinic acid, monomercaptosuccinic acid, butanedioic acid, mercapto, malic acid, 2-thio, mercaptosuccinate, 2-sulfanylsuccinic acid, dl-mercaptosuccinic acid | |
| CHEBI:38705 | |
| C(C(C(=O)O)S)C(=O)O |
Safety and Handling
EINECSNumber : (2)-1451
RTECSNumber : WM8225000
TSCA : Yes