missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiolactic Acid 97.0+%, TCI America™

Supplier:  TCI America M006025G

 View more versions of this product

Catalog No. M006025G



Description

Description

Specifications

Specifications

Thiolactic Acid
Colorless
C3H6O2S
25 g
2-mercaptopropionic acid, thiolactic acid, 2-mercaptopropanoic acid, 2-thiolactic acid, 2-thiolpropionic acid, alpha-mercaptopropionic acid, alpha-mercaptopropanoic acid, propionic acid, 2-mercapto, propanoic acid, 2-mercapto, fema no. 3180
CC(C(=O)O)S
106.139
CHEBI:47872
≥97.0% (GC)
79-42-5
115°C
MFCD00004862
2936
PMNLUUOXGOOLSP-UHFFFAOYSA-N
2-sulfanylpropanoic acid
62326
106.14
Liquid

Chemical Identifiers

79-42-5
106.139
PMNLUUOXGOOLSP-UHFFFAOYSA-N
62326
2-sulfanylpropanoic acid
C3H6O2S
MFCD00004862
2-mercaptopropionic acid, thiolactic acid, 2-mercaptopropanoic acid, 2-thiolactic acid, 2-thiolpropionic acid, alpha-mercaptopropionic acid, alpha-mercaptopropanoic acid, propionic acid, 2-mercapto, propanoic acid, 2-mercapto, fema no. 3180
CHEBI:47872
CC(C(=O)O)S
Safety and Handling

Safety and Handling

RTECSNumber : UF5250000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions