Please login to your online account to display your discounted pricing

Alfa Aesar™ Thioisonicotinamide, 98%

$45.20 - $373.68

Chemical Identifiers

Chemical Name or Material Thioisonicotinamide
CAS 2196-13-6
Molecular Formula C6H6N2S
Formula Weight 138.19
MDL Number MFCD00006437
Beilstein 2174
Synonym Pyridine-4-thiocarboxamide; Isothionicotinamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2526503

  SDS
alfa aesar™
B2526503
1g Each for $45.20

AAB2526506

  SDS
alfa aesar™
B2526506
5g Each for $139.68

AAB2526514

  SDS
alfa aesar™
B2526514
25g Each for $373.68
Description & Specifications

Specifications

Formula Weight 138.19
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym Pyridine-4-thiocarboxamide; Isothionicotinamide
Health Hazard 1 Warning
Melting Point 198° to 202°C (dec.)
Chemical Name or Material Thioisonicotinamide
Assay 98%
Molecular Formula C6H6N2S
TSCA TSCA
MDL Number MFCD00006437
Beilstein 2174
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 2196-13-6
Quantity 1g
RTECS Number US4204000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.