missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thioglycolic Acid 98.0+%, TCI America™
Supplier: TCI America M005225G
Specifications
Thioglycolic Acid | |
-16°C | |
96°C | |
MFCD00004876 | |
1940 | |
CWERGRDVMFNCDR-UHFFFAOYSA-N | |
2-sulfanylacetic acid | |
1133 | |
92.11 | |
Liquid |
68-11-1 | |
Colorless | |
C2H4O2S | |
25 g | |
mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio | |
C(C(=O)O)S | |
92.112 | |
CHEBI:30065 | |
≥98.0% (T) |
Chemical Identifiers
68-11-1 | |
92.112 | |
CWERGRDVMFNCDR-UHFFFAOYSA-N | |
1133 | |
2-sulfanylacetic acid |
C2H4O2S | |
MFCD00004876 | |
mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio | |
CHEBI:30065 | |
C(C(=O)O)S |
Safety and Handling
EINECSNumber : (2)-1355
RTECSNumber : AI5950000
TSCA : Yes
Recommended Storage : Refrigerator