missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thioglycolic acid, 99.94%, Bacteriological grade, MP Biomedicals™

Manufacturer:  Mp Biomedicals 10293325

 View more versions of this product

Catalog No. ICN10293325



Specifications

Specifications

Thioglycolic acid
White
367-51-1
erhavit d, mercaptoacetic acid sodium salt, mercaptoacetic acid, sodium salt, mollescal sf, sodium 2-mercatoethanoate, sodium 2-sulfanylacetate, sodium mercaptoacetate, sodium thioglycolate, sodium thioglycollate, thioglycolate sodium
C(C(=O)[O-])S.[Na+]
114.094
CHEBI:86481
Crystalline Powder
Bacteriological
∽1.32g/mL
25g
C2H3NaO2S
GNBVPFITFYNRCN-UHFFFAOYSA-M
sodium;2-sulfanylacetate
23690444
114.11
0.9994

Chemical Identifiers

367-51-1
114.094
erhavit d, mercaptoacetic acid sodium salt, mercaptoacetic acid, sodium salt, mollescal sf, sodium 2-mercatoethanoate, sodium 2-sulfanylacetate, sodium mercaptoacetate, sodium thioglycolate, sodium thioglycollate, thioglycolate sodium
CHEBI:86481
C(C(=O)[O-])S.[Na+]
C2H3NaO2S
GNBVPFITFYNRCN-UHFFFAOYSA-M
23690444
sodium;2-sulfanylacetate
Documents