missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mercaptoacetic acid, 97+%, Thermo Scientific Chemicals

Air Senstitive

Supplier:  Thermo Scientific Chemicals B2039136

 View more versions of this product

Catalog No. AAB2039136



Description

Description

Reagent that protects tryptophan in amino acid analysis.Mercaptoacetic acid is used as a protecting agent for tryptophan in amino acid analysis and an acidity indicator. It finds application as an intermediate in the chemical reactions such as addition, elimination and cyclization. It acts as a precursor to ammonium thioglycolate, sodium thioglycolate and calcium thioglycolate. Its organotin derivatives are used as stabilizers for polyvinyl chloride (PVC). In organic synthesis, it acts as a nucleophile in thioglycolysis reactions and sulfur transfer agent for sulfonyl chloride synthesis. Further, it is used in leather processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent that protects tryptophan in amino acid analysis.Mercaptoacetic acid is used as a protecting agent for tryptophan in amino acid analysis and an acidity indicator. It finds application as an intermediate in the chemical reactions such as addition, elimination and cyclization. It acts as a precursor to ammonium thioglycolate, sodium thioglycolate and calcium thioglycolate. Its organotin derivatives are used as stabilizers for polyvinyl chloride (PVC). In organic synthesis, it acts as a nucleophile in thioglycolysis reactions and sulfur transfer agent for sulfonyl chloride synthesis. Further, it is used in leather processing.

Solubility
Miscible with water, ethanol, ethers, ketones, esters, chlorinated hydrocarbons, benzene and aromatic hydrocarbons. Slightly miscible with chloroform.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

Mercaptoacetic acid
68-11-1
1.295
119°C (246°F)
1.5045
MFCD00004876
UN1940
Air Sensitive
mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio
CWERGRDVMFNCDR-UHFFFAOYSA-N
2-sulfanylacetic acid
1133
92.11
Liquid
98%
°C to 12°C
220°C
C2H4O2S
HSCH2CO2H
500 g
506166
14,9336
Miscible with water,ethanol,ethers,ketones,esters,chlorinated hydrocarbons,benzene and aromatic hydrocarbons. Slightly miscible with chloroform.
C(C(=O)O)S
92.112
CHEBI:30065
≥97%

Chemical Identifiers

68-11-1
92.112
CWERGRDVMFNCDR-UHFFFAOYSA-N
1133
2-sulfanylacetic acid
C2H4O2S
MFCD00004876
mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio
CHEBI:30065
C(C(=O)O)S
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H314

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H311+H331-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: THIOGLYCOLIC ACID

EINECSNumber : 200-677-4

RTECSNumber : AI5950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only