missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thioflavin T, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 211761000

 View more versions of this product

Catalog No. AC211761000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Thioflavin T
C17H19ClN2S
27, 377
Solubility in water: soluble. Other solubilities: soluble in most organic solvent
Authentic
2-[4-(dimethylamino)phenyl]-3,6-dimethyl-1,3-benzothiazol-3-ium chloride
16953
318.85
Complies
100 g
2390-54-7
MFCD00011944
Basic Yellow 1, C.I. 49005
JADVWWSKYZXRGX-UHFFFAOYSA-M
[Cl-].CN(C)C1=CC=C(C=C1)C1=[N+](C)C2=CC=C(C)C=C2S1
318.86
CHEBI:76023
Glass bottle
Ochre/Yellow to Yellow
Powder

Chemical Identifiers

2390-54-7
318.86
JADVWWSKYZXRGX-UHFFFAOYSA-M
16953
2-[4-(dimethylamino)phenyl]-3,6-dimethyl-1,3-benzothiazol-3-ium chloride
C17H19ClN2S
MFCD00011944
Basic Yellow 1, C.I. 49005
CHEBI:76023
[Cl-].CN(C)C1=CC=C(C=C1)C1=[N+](C)C2=CC=C(C)C=C2S1
Safety and Handling

Safety and Handling

EINECSNumber : 219-228-9

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only