Please login to your online account to display your discounted pricing

Thioflavin T, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC211760050

 View more versions of this product

Catalog No. AC211760050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 318.85
Quantity 5g
Color Yellow to ochre-yellow
Beilstein 27, 377
TSCA TSCA
EINECS Number 219-228-9
Chemical Name or Material Thioflavin T
Molecular Formula C17H19ClN2S
MDL Number MFCD00011944
CAS 2390-54-7
Synonym Basic Yellow 1
Infrared Spectrum Authentic
Packaging Glass bottle
UV Spectrometry Complies

Specifications

Chemical Name or Material Thioflavin T
CAS 2390-54-7
Molecular Formula C17H19ClN2S
Formula Weight 318.85
MDL Number MFCD00011944
Synonym Basic Yellow 1