Please login to your online account to display your discounted pricing

Alfa Aesar™ Thiodisuccinic acid, 98+%

$41.58 - $125.07

Chemical Identifiers

Chemical Name or Material Thiodisuccinic acid
CAS 4917-76-4
Molecular Formula C8H10O8S
Formula Weight 266.22
MDL Number MFCD00020502
Beilstein 1804352
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0322206

  SDS
alfa aesar™
L0322206
5g Each for $41.58

AAL0322214

  SDS
alfa aesar™
L0322214
25g Each for $125.07
Description & Specifications

Specifications

Formula Weight 266.22
MDL Number MFCD00020502
Molecular Formula C8H10O8S
Chemical Name or Material Thiodisuccinic acid
CAS 4917-76-4
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 5g
Melting Point ≈208°C
Beilstein 1804352
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.