missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiocarbohydrazide, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 207531000

Catalog No. AC207531000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Thiocarbohydrazide
2231-57-4
97.5% min
CH6N4S
MFCD00007616
03, 197
LJTFFORYSFGNCT-UHFFFAOYSA-N
1,3-diaminothiourea
2724189
106.15
98%
Authentic
Plastic bottle
H2NNHCSNHNH2
100g
thiocarbohydrazide, thiocarbazide, thiocarbonohydrazide, carbonothioic dihydrazide, carbohydrazide, thio, thiocarbonic dihydrazide, 1,3-diamino-2-thiourea, hydrazinecarbohydrazonothioic acid, usaf ek-7372, unii-1iz2h82nwu
C(=S)(NN)NN
106.15
CHEBI:9549
98%
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Fatal if inhaled.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat,drink or smoke when using th

GHS Signal Word: Danger

EINECSNumber : 218-769-8

RTECSNumber : FF2975000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only