Please login to your online account to display your discounted pricing

Alfa Aesar™ Thiobenzoic acid, 94%

$50.03 - $137.26

Chemical Identifiers

Chemical Name or Material Thiobenzoic acid
CAS 98-91-9
Molecular Formula C7H6OS
Formula Weight 138.19
MDL Number MFCD00004852
Beilstein 1071790
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2445822

  SDS
alfa aesar™
B2445822
100g Each for $50.03

AAB2445836

  SDS
alfa aesar™
B2445836
500g Each for $137.26
Description & Specifications

Specifications

Formula Weight 138.19
Melting Point 15° to 18°C
Boiling Point 103° to 105°C (12mmHg)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage 2° to 8°C
RTECS Number DH6839000
Flash Point >110°C (230°F)
Molecular Formula C7H6OS
CAS 98-91-9
Chemical Name or Material Thiobenzoic acid
Quantity 100g
Beilstein 1071790
TSCA TSCA
MDL Number MFCD00004852
Assay 94%
Health Hazard 1 Warning
Odor Stench

Applications
Thiobenzoic acid is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C