Please login to your online account to display your discounted pricing

Thiobenzoic acid, 90%, ACROS Organics™

$25.64 - $25.64

Chemical Identifiers

Chemical Name or Material Thiobenzoic acid
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C7H6OS
Linear Formula C6H5COSH
Formula Weight 138.18
MDL Number MFCD00004852
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC138685000

  SDS
thermo fisher scientific
AC138685000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC138680250

  SDS
thermo fisher scientific
138680250
Glass bottle 25g Each for $25.64

AC138681000

  SDS
thermo fisher scientific
138681000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥88%
Formula Weight 138.18
Quantity 500g
Color Clear yellow to brown
Density 1.17g/mL
Beilstein 09, 419
Linear Formula C6H5COSH
Infrared Spectrum Authentic
Refractive Index 1.6020 to 1.6080 (20°C, 589nm)
Molecular Formula C7H6OS
Chemical Name or Material Thiobenzoic acid
CAS 98-91-9
Assay Percent Range 90%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00004852
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.