missing translation for 'onlineSavingsMsg'
Learn More

Thioacetic Acid 95.0+%, TCI America™

Supplier:  TCI America T0189500ML

 View more versions of this product

Catalog No. T0189500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Thioacetic Acid
Yellow
C2H4OS
500 mL
thioacetic acid, ethanethioic acid, thiacetic acid, thioacetic s-acid, acetyl mercaptan, ethanethiolic acid, thiolacetic acid, thionoacetic acid, methanecarbothiolic acid, ch3cosh
CC(S)=O
76.11
CHEBI:16555
≥95.0% (GC)
507-09-5
93°C
MFCD00004853
2436
DUYAAUVXQSMXQP-UHFFFAOYSA-N
ethanethioic S-acid
10484
76.11
Liquid

Chemical Identifiers

507-09-5
76.11
DUYAAUVXQSMXQP-UHFFFAOYSA-N
10484
ethanethioic S-acid
C2H4OS
MFCD00004853
thioacetic acid, ethanethioic acid, thiacetic acid, thioacetic s-acid, acetyl mercaptan, ethanethiolic acid, thiolacetic acid, thionoacetic acid, methanecarbothiolic acid, ch3cosh
CHEBI:16555
CC(S)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0691

RTECSNumber : AJ5600000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions