Please login to your online account to display your discounted pricing

Thioacetamide, ACS Grade, LabChem™

$259.26 - $259.26

Chemical Identifiers

Chemical Name or Material Thioacetamide
CAS 62-55-5
CAS Max % 100
Purity Grade ACS Grade
Linear Formula CH3CSNH2
Formula Weight 75.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC260009

 
lab chem
LC260009
Amber Glass 100g Each for $259.26
Description & Specifications

Specifications

Formula Weight 75.13
Identification Passes Test
Melting Point 113°C
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Storage Information Room Temperature
Color White to off-white
Solubility Soluble in water
Chemical Name or Material Thioacetamide
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause cancer.
Harmful to aquatic life with long lasting effects.
Linear Formula CH3CSNH2
CAS 62-55-5
Physical Form Solid
Packaging Amber Glass
EINECS Number 200-541-4
Quantity 100g
Purity Grade ACS Grade
Health Hazard 1 Danger
CAS Max % 100
Safety and Handling

Danger

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause cancer.
Harmful to aquatic life with long lasting effects.