Learn More
Thioacetamide, ACS, 99% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03063809
Description
Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Thioacetamide | |
112°C to 114°C | |
MFCD00008070 | |
506006 | |
thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide | |
YUKQRDCYNOVPGJ-UHFFFAOYSA-N | |
ethanethioamide | |
2723949 | |
75.13 | |
ACS |
62-55-5 | |
C2H5NS | |
10 g | |
14,9319 | |
Soluble in ethanol,benzene and petroleum ether. Slightly soluble in water and ether. | |
CC(N)=S | |
75.13 | |
CHEBI:32497 | |
99% | |
Crystalline |
Chemical Identifiers
62-55-5 | |
75.13 | |
YUKQRDCYNOVPGJ-UHFFFAOYSA-N | |
2723949 | |
ethanethioamide |
C2H5NS | |
MFCD00008070 | |
thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide | |
CHEBI:32497 | |
CC(N)=S |
Safety and Handling
GHS H Statement
H350-H302-H315-H319
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H319-H336-H351-H373
EINECSNumber : 200-541-4
RTECSNumber : AC8925000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only