missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Thioacetamide, ACS, 99%

Manufacturer:  Alfa Aesar™ 3063818

 View more versions of this product

Catalog No. AA3063818



Specifications

Specifications

Thioacetamide
50g
ACS Grade
C2H5NS
506006
Ethanethioamide
CC(=S)N
75.129
CHEBI:32497
Crystalline
112°C to 114°C
62-55-5
99%
MFCD00008070
14,9319
YUKQRDCYNOVPGJ-UHFFFAOYSA-N
ethanethioamide
2723949
75.13

Chemical Identifiers

62-55-5
75.129
YUKQRDCYNOVPGJ-UHFFFAOYSA-N
2723949
ethanethioamide
C2H5NS
MFCD00008070
Ethanethioamide
CHEBI:32497
CC(=S)N
Safety and Handling

Safety and Handling

GHS H Statement
H350-H302-H315-H319
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P281-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : AC8925000

TSCA : TSCA

SDS Documents