Epredia™ Thinline Biopsy Cassettes in Tube Packs
For biopsy tissue samples, cassettes have a front lid hinge, attached lids in matching colors, and are compatible with all Epredia PrintMate™ Cassette Printers
$1,253.47 - $1,253.47
Specifications
Closure Included | Yes |
---|---|
Closure Type | Attached Lids, Front Hinge |
Description | For Use With PrintMate™ Printers |
Item Description | Thinline Biopsy Cassettes |
Packaging Type | 75/Pk., 10 Pk./Cs |
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
B851737BE
|
Epredia™
B851737BE |
Beige |
Case of 750 Each for $1,253.47
|
|
B851737BL
|
Epredia™
B851737BL |
Blue |
Case of 750 Each for $1,253.47
|
|
B851737GN
|
Epredia™
B851737GN |
Green |
Case of 750 Each for $1,253.47
|
|
B851737GY
|
Epredia™
B851737GY |
Gray |
Case of 750 Each for $1,253.47
|
|
B851737LA
|
Epredia™
B851737LA |
Lavender |
Case of 750 Each for $1,253.47
|
|
B851737LG
|
Epredia™
B851737LG |
Light Green |
Case of 750 Each for $1,253.47
|
|
B851737LI
|
Epredia™
B851737LI |
Lilac |
Case of 750 Each for $1,253.47
|
|
B851737OR
|
Epredia™
B851737OR |
Orange |
Case of 750 Each for $1,253.47
|
|
B851737TN
|
Epredia™
B851737TN |
Tan |
Case of 750 Each for $1,253.47
|
|
B851737WH
|
Epredia™
B851737WH |
White |
Case of 750 Each for $1,253.47
|
|
B851737YW
|
Epredia™
B851737YW |
Yellow |
Case of 750 Each for $1,253.47
|
|
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Yes | |
For Use With PrintMate™ Printers | |
75/Pk., 10 Pk./Cs | |
Yes | |
Biopsy Cassette |
Attached Lids, Front Hinge | |
Thinline Biopsy Cassettes | |
Small Squares | |
750/Cs |