Epredia™ Thinline Biopsy Cassettes in Tube Packs
For biopsy tissue samples, cassettes have a front lid hinge, attached lids in matching colors, and are compatible with all Epredia PrintMate™ Cassette Printers
Manufacturer: Epredia™ B851737LI
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Yes | |
Small Squares | |
Biopsy Cassette | |
Yes | |
For Use With PrintMate™ Printers |
75/Pk., 10 Pk./Cs | |
750/Cs | |
Attached Lids, Front Hinge | |
Lilac | |
Thinline Biopsy Cassettes |