missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde, TCI America™

Manufacturer:  TCI America T32121G

 View more versions of this product

Catalog No. T32121G



Specifications

Specifications

Thieno[3,2-b]thiophene-2,5-dicarboxaldehyde
274°C
37882-75-0
MFCD03931117
C1=C(SC2=C1SC(=C2)C=O)C=O
196.238
196.24
≥93.0% (GC)
Undesignated
1g
C8H4O2S2
UBKNFAFBINVTOP-UHFFFAOYSA-N
thieno[3,2-b]thiophene-2,5-dicarbaldehyde
361865
Crystal-Powder at 20°C

Chemical Identifiers

37882-75-0
196.238
UBKNFAFBINVTOP-UHFFFAOYSA-N
thieno[3,2-b]thiophene-2,5-dicarbaldehyde
C8H4O2S2
MFCD03931117
361865
C1=C(SC2=C1SC(=C2)C=O)C=O
Safety and Handling

Safety and Handling

TSCA : No

Documents