missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thieno[3,2-b]pyridine-2-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 347187-30-8
Molecular Formula C8H5NO2S
Molecular Weight (g/mol) 179.193
MDL Number MFCD12198115
InChI Key WVWCTPQHJWMLKI-UHFFFAOYSA-N
Synonym thieno 3,2-b pyridine-2-carboxylic acid, thiopheno 3,2-b pyridine-2-carboxylic acid
PubChem CID 22047169
IUPAC Name thieno[3,2-b]pyridine-2-carboxylic acid
SMILES C1=CC2=C(C=C(S2)C(=O)O)N=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC70301CB
SDS View Documents Promotion Details
Fisher Scientific Company
CC70301CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC70301DA
SDS View Documents Promotion Details
Fisher Scientific Company
CC70301DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

347187-30-8
179.193
WVWCTPQHJWMLKI-UHFFFAOYSA-N
22047169
C1=CC2=C(C=C(S2)C(=O)O)N=C1
C8H5NO2S
MFCD12198115
thieno 3,2-b pyridine-2-carboxylic acid, thiopheno 3,2-b pyridine-2-carboxylic acid
thieno[3,2-b]pyridine-2-carboxylic acid

Specifications

347187-30-8
100.0
C8H5NO2S
Amber Glass Bottle
250mg
C1=CC2=C(C=C(S2)C(=O)O)N=C1
179.193
179.2
Thieno[3, 2-b]pyridine-2-carboxylic acid
96.5
97%
MFCD12198115
thieno 3,2-b pyridine-2-carboxylic acid, thiopheno 3,2-b pyridine-2-carboxylic acid
WVWCTPQHJWMLKI-UHFFFAOYSA-N
thieno[3,2-b]pyridine-2-carboxylic acid
22047169
97%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Product Certifications