missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thieno[2,3-b]pyrazine-6-carboxylic acid, 90%, Maybridge

Chemical Identifiers

Assay Percent Range 90%
Molecular Formula C7H4N2O2S
Formula Weight 180.19
Chemical Name or Material Thieno[2, 3-b]pyrazine-6-carboxylic acid
Assay 90%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC31501DA SDS Fisher Scientific Company
CC31501DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC31501CB SDS Fisher Scientific Company
CC31501CB
Amber Glass Bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

90%
180.19
90%
C7H4N2O2S
Thieno[2, 3-b]pyrazine-6-carboxylic acid

Specifications

180.19
59944-79-5
Amber Glass Bottle
Thieno[2, 3-b]pyrazine-6-carboxylic acid
1g
90%
C7H4N2O2S
90%