Please login to your online account to display your discounted pricing

Thieno[2,3-b]pyrazine-6-carboxylic acid, 90%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC31501DA

 View more versions of this product

Catalog No. CC31501DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 180.19
Quantity 1g
Assay 90%
Chemical Name or Material Thieno[2, 3-b]pyrazine-6-carboxylic acid
Assay Percent Range 90%
CAS 59944-79-5
Packaging Amber Glass Bottle
Molecular Formula C7H4N2O2S

Specifications

Chemical Name or Material Thieno[2, 3-b]pyrazine-6-carboxylic acid
Assay 90%
Assay Percent Range 90%
Molecular Formula C7H4N2O2S
Formula Weight 180.19