Please login to your online account to display your discounted pricing

Thiazolidine-2-carboxylic acid, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  353020010

 View more versions of this product

Catalog No. AC353020010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 133.17
Quantity 1g
Color White
Melting Point 179° to 184°C
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00068005
CAS 16310-13-7
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Thiazolidine-2-carboxylic acid
Name Note 97%
Assay Percent Range 97%
Molecular Formula C4 H7 N O2 S

Specifications

Chemical Name or Material Thiazolidine-2-carboxylic acid
Name Note 97%
CAS 16310-13-7
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4 H7 N O2 S
Formula Weight 133.17
MDL Number MFCD00068005
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents