Please login to your online account to display your discounted pricing

Thiazolidine-2-carboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Thiazolidine-2-carboxylic acid
Name Note 97%
CAS 16310-13-7
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4 H7 N O2 S
Formula Weight 133.17
MDL Number MFCD00068005
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC353020010

  SDS
thermo fisher scientific
353020010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 133.17
Physical Form Powder
Quantity 1g
Color White
Melting Point 179° to 184°C
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 16310-13-7
MDL Number MFCD00068005
Name Note 97%
Chemical Name or Material Thiazolidine-2-carboxylic acid
Molecular Formula C4 H7 N O2 S
Assay Percent Range 97%
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.