Learn More
Thiazole-2-carboxaldehyde, 95%, Thermo Scientific Chemicals
$80.62 - $301.73
Chemical Identifiers
CAS | 10200-59-6 |
---|---|
Molecular Formula | C4H3NOS |
Molecular Weight (g/mol) | 113.134 |
MDL Number | MFCD00142924 |
InChI Key | ZGTFNNUASMWGTM-UHFFFAOYSA-N |
Synonym | 2-thiazolecarboxaldehyde, thiazole-2-carbaldehyde, 2-formylthiazole, thiazole-2-carboxaldehyde, 2-formyl thiazole, thiazolecarboxaldehyde, 2-formyl-1,3-thiazole, 1,3-thiazole-2-carboxaldehyde, 2-thiazolecarbaldehyde, pubchem9929 |
PubChem CID | 2734903 |
ChEBI | CHEBI:43623 |
IUPAC Name | 1,3-thiazole-2-carbaldehyde |
SMILES | C1=CSC(=N1)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6332903
|
Thermo Scientific Chemicals
H6332903 |
1 g |
Each for $80.62
|
|
|||||
AAH6332906
|
Thermo Scientific Chemicals
H6332906 |
5 g |
Each for $301.73
|
|
|||||
Description
Thiazole-2-carboxaldehyde may be used as a reactant in the following syntheses Benzothiazine N-acylhydrazones, having potential antinociceptive and anti-inflammatory activity, Thiazole-2-yl-(amino)methylphosphonate diethyl esters, Imino ester by reacting with L-leucine t-butyl ester hydrochloride. Also used in building block for taxane analogs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThiazole-2-carboxaldehyde may be used as a reactant in the following syntheses Benzothiazine N-acylhydrazones, having potential antinociceptive and anti-inflammatory activity, Thiazole-2-yl-(amino)methylphosphonate diethyl esters, Imino ester by reacting with L-leucine t-butyl ester hydrochloride. Also used in building block for taxane analogs.
Solubility
Fully miscible with water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
10200-59-6 | |
113.134 | |
ZGTFNNUASMWGTM-UHFFFAOYSA-N | |
2734903 | |
1,3-thiazole-2-carbaldehyde |
C4H3NOS | |
MFCD00142924 | |
2-thiazolecarboxaldehyde, thiazole-2-carbaldehyde, 2-formylthiazole, thiazole-2-carboxaldehyde, 2-formyl thiazole, thiazolecarboxaldehyde, 2-formyl-1,3-thiazole, 1,3-thiazole-2-carboxaldehyde, 2-thiazolecarbaldehyde, pubchem9929 | |
CHEBI:43623 | |
C1=CSC(=N1)C=O |
Specifications
10200-59-6 | |
1.288 g/mL | |
68°C (154°F) | |
1.574 | |
1 g | |
Fully miscible with water. | |
C1=CSC(=N1)C=O | |
113.134 | |
CHEBI:43623 | |
95% | |
Thiazole-2-carboxaldehyde |
Colorless to Yellow | |
61°C to 63°C (15 mmHg) | |
C4H3NOS | |
MFCD00142924 | |
2-thiazolecarboxaldehyde, thiazole-2-carbaldehyde, 2-formylthiazole, thiazole-2-carboxaldehyde, 2-formyl thiazole, thiazolecarboxaldehyde, 2-formyl-1,3-thiazole, 1,3-thiazole-2-carboxaldehyde, 2-thiazolecarbaldehyde, pubchem9929 | |
ZGTFNNUASMWGTM-UHFFFAOYSA-N | |
1,3-thiazole-2-carbaldehyde | |
2734903 | |
113.13 | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only