missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thianaphthene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 138620250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Thianaphthene | |
28.0°C to 32.0°C | |
1.1400g/mL | |
110°C | |
96% min. (GC) | |
C8H6S | |
25 g | |
1.14 | |
benzo b thiophene, thianaphthene, benzothiophene, benzothiofuran, thionaphthene, 1-thiaindene, thianaphthen, benzothiophen, thianaphtene, 1-benzothiophen | |
FCEHBMOGCRZNNI-UHFFFAOYSA-N | |
1-benzothiophene | |
7221 | |
134.2 | |
Crystalline Solid |
95-15-8 | |
Pink to White or Yellow | |
221.0°C to 222.0°C | |
Authentic | |
Glass bottle | |
MFCD00005864 | |
17, 59 | |
15, 9455 | |
Solubility in water: insoluble. Other solubilities: soluble in usual organic solvents, soluble in ethanol, ethylether, acetone | |
C1=CC=C2C(=C1)C=CS2 | |
134.2 | |
CHEBI:35858 | |
97% |
Chemical Identifiers
95-15-8 | |
134.2 | |
FCEHBMOGCRZNNI-UHFFFAOYSA-N | |
7221 | |
1-benzothiophene |
C8H6S | |
MFCD00005864 | |
benzo b thiophene, thianaphthene, benzothiophene, benzothiofuran, thionaphthene, 1-thiaindene, thianaphthen, benzothiophen, thianaphtene, 1-benzothiophen | |
CHEBI:35858 | |
C1=CC=C2C(=C1)C=CS2 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Warning
EINECSNumber : 202-395-7
RUO – Research Use Only