Please login to your online account to display your discounted pricing

Thian-4-one, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Thian-4-one
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H8OS
Formula Weight 116.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF03869DA

 
fisher scientific company
RF03869DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 116.18
Quantity 1g
Assay 97%
Chemical Name or Material Thian-4-one
Assay Percent Range 97%
Molecular Formula C5H8OS
CAS 1072-72-6