Please login to your online account to display your discounted pricing

Thian-4-one, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  RF03869DA

 View more versions of this product

Catalog No. RF03869DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 116.18
Quantity 1g
Assay 97%
Chemical Name or Material Thian-4-one
Assay Percent Range 97%
Molecular Formula C5H8OS
CAS 1072-72-6

Specifications

Chemical Name or Material Thian-4-one
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H8OS
Formula Weight 116.18